Skip to Content
Merck

Skip To

A5028

Adenylosuccinic acid

~96% (HPLC)

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C14H18N5O11P
CAS Number:
Molecular Weight:
463.29
PubChem Substance ID:
UNSPSC Code:
12352204


assay

~96% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP(O)(O)=O)n2cnc3c(N[C@@H](CC(O)=O)C(O)=O)ncnc23

InChI

1S/C14H18N5O11P/c20-7(21)1-5(14(24)25)18-11-8-12(16-3-15-11)19(4-17-8)13-10(23)9(22)6(30-13)2-29-31(26,27)28/h3-6,9-10,13,22-23H,1-2H2,(H,20,21)(H,24,25)(H,15,16,18)(H2,26,27,28)/t5-,6+,9+,10+,13+/m0/s1

InChI key

OFBHPPMPBOJXRT-VWJPMABRSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



B Rees et al.
Journal of molecular biology, 299(5), 1157-1164 (2000-06-30)
The crystal structure of aspartyl-tRNA synthetase from Escherichia coli has been determined to a resolution of 2.7 A. The structure is compared to the same enzyme co-crystallized with tRNA(Asp) and containing aspartyl adenylate or ATP. The asymmetric unit contains three
Maria Novikova et al.
The Journal of biological chemistry, 285(17), 12662-12669 (2010-02-18)
The heptapeptide-nucleotide microcin C (McC) is a potent inhibitor of enteric bacteria growth. McC is excreted from producing cells by the MccC transporter. The residual McC that remains in the producing cell can be processed by cellular aminopeptidases with the
H Mejdoub et al.
Biochemistry, 26(7), 2054-2059 (1987-04-07)
Aspartyl-tRNA synthetase from bakers' yeast gives an unstable complex with the cognate adenylate, which reacts after dissociation with amino acid side chains of the protein. This leads to a covalent incorporation of aspartic acid into aspartyl-tRNA synthetase via amide or