Skip to Content
Merck

Skip To

A4302

L-Alanine 7-amido-4-methylcoumarin trifluoroacetate salt

aminopeptidase substrate, fluorogenic, ≥98% (TLC), powder

Synonym(s):

L-Alanine 4-methyl-7-coumarinylamide trifluoroacetate salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 mg
Check Cart for Availability
€ 128,00
250 mg
Check Cart for Availability
€ 562,00

About This Item

Empirical Formula (Hill Notation):
C13H14N2O3 · C2HF3O2
CAS Number:
Molecular Weight:
360.29
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83
MDL number:

€ 128,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Alanine 7-amido-4-methylcoumarin trifluoroacetate salt, aminopeptidase substrate

Quality Segment

assay

≥98% (TLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless to very faintly yellow

fluorescence

λex 325 nm; λem 389 nm in ethanol

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.C[C@H](N)C(=O)Nc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C13H14N2O3.C2HF3O2/c1-7-5-12(16)18-11-6-9(3-4-10(7)11)15-13(17)8(2)14;3-2(4,5)1(6)7/h3-6,8H,14H2,1-2H3,(H,15,17);(H,6,7)/t8-;/m0./s1

InChI key

YYGKKBUKGNFDJW-QRPNPIFTSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A2027A2628C8022
assay

≥98% (TLC)

assay

≥97% (TLC)

assay

≥98% (TLC)

assay

≥98% (TLC)

solubility

water: 50 mg/mL, clear, colorless to very faintly yellow

solubility

acetic acid: water (1:1): 50 mg/mL, clear, colorless to light yellow

solubility

ethanol: 50 mg/mL, clear to slightly hazy

solubility

methanol: 20 mg/mL, clear, colorless to faintly yellow

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

fluorescence

λex 325 nm; λem 389 nm in ethanol

fluorescence

-

fluorescence

-

fluorescence

λex 290 nm; λem 390 nm (buffer pH 7.1), λex 290 nm; λem 437 nm (Reaction product)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
A4302-250MG04061833054673
A4302-25MG04061833054680

Questions

1–2 of 2 Questions  
  1. Can Product A4302, L-Alanine 7-amido-4-methylcoumarin trifluoroacetate salt be utilized as a substrate for Aminopeptidase?

    1 answer
    1. The use of L-Alanine 7-Amido-4-methylcoumarin as a substrate for Aminopeptidase is described in Biochem. J., 211, 567 - 573 (1982). The excitaion and emission maximum of the liberated NH2-Mec is 380 nm and 440 nm respectively.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters