Skip to Content
Merck
All Photos(1)

Key Documents

A3355

Sigma-Aldrich

ATC0175

≥98% (HPLC), solid

Synonym(s):

N-(cis-4-{[4-(Dimethylamino)quinazolin-2-yl]amino}cyclohexyl)-3,4-difluorobenzamide HCl

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€345.00

€345.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€345.00

About This Item

Empirical Formula (Hill Notation):
C23H26ClF2N5O
CAS Number:
Molecular Weight:
461.94
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

€345.00


Estimated to ship on26 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: soluble 5 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

Cl[H].CN(C)c1nc(N[C@@H]2CC[C@@H](CC2)NC(=O)c3ccc(F)c(F)c3)nc4ccccc14

InChI

1S/C23H25F2N5O.ClH/c1-30(2)21-17-5-3-4-6-20(17)28-23(29-21)27-16-10-8-15(9-11-16)26-22(31)14-7-12-18(24)19(25)13-14;/h3-7,12-13,15-16H,8-11H2,1-2H3,(H,26,31)(H,27,28,29);1H/t15-,16+;

InChI key

HUUPKFUYSQNNLO-FAESNJTISA-N

Biochem/physiol Actions

ATC0175 is a non-peptide, orally active melanin-concentrating hormone receptor 1 (MCHR1) antagonist.
Non-peptide, orally active melanin-concentrating hormone receptor 1 (MCHR1) antagonist.

Features and Benefits

This compound is featured on the Melanin-Concentrating Hormone Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigeyuki Chaki et al.
The Journal of pharmacology and experimental therapeutics, 313(2), 831-839 (2005-01-29)
Melanin-concentrating hormone (MCH) is a cyclic peptide produced in the lateral hypothalamus. It has been implicated in a number of physiological processes including feeding behavior, energy balance, and the regulation of emotional states. Here, we report in vitro and in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service