Skip to Content
Merck

Skip To

A3188

Anti-Human IgG (γ-chain specific)−Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous glycerol solution

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
MDL number:
Conjugate:
alkaline phosphatase conjugate
Clone:
polyclonal
Application:
ELISA (d), IHC (p), WB
Citations:
9


biological source

goat

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

should not react with

mouse

technique(s)

direct ELISA: 1:30,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50, western blot: 1:30,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Human IgG antibodies are primarily responsible for the facilitation of humoral immune responses such as complement fixation, placental transport, and phagocytosis. Anti-human IgG (γ-chain specific) −alkaline phosphatase antibody can be used in immunohistochemistry and immunoelectrophoresis. It can also be used in western blotting. Goat anti-human IgG−alkaline phosphatase antibody reacts specifically with human IgG when tested against human IgA, IgG, IgM, Bence Jones κ, and λ myeloma proteins. The product is not reactive with mouse IgG or mouse serum proteins.

Immunogen

Purified human IgG.

Application

Anti-human IgG (γ-chain specific) −alkaline phosphatase antibody can be used in immunocytochemical staining techniques. It can also be used for dot blot.
Proteins harvested from E. coli cultures were run on SDS-page gel for western blot analysis using alkaline phosphatase conjugated goat anti-human IgG (chain specific) at a concentration of 1:10000 for the secondary. Membranes were developed using BCIP/NBT substrate (Sigma).[1]
Proteins harvested from E. coli cultures were run on SDS-page gel for western blot analysis using alkaline phosphatase conjugated goat anti-human IgG (chain specific) at a concentration of 1:10000 for the secondary.[1] IgG was detected in human serum by Elisa using alkaline phosphatase conjugated goat anti-human IgG (gamma-chain specific) diluted 1:5000 in 0.25% BSA/PBS. Assay was developed using p-nitrophenyl phosphate substrate (1mg/ml Sigma).[2]

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 10 mM glycine, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide as preservative

Other Notes

Antibody adsorbed with mouse serum proteins.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions






Global Trade Item Number

SKUGTIN
A3188-.5ML04061835804979
A3188-.25ML04061835804962

Questions

Reviews

No rating value

Active Filters