Skip to Content
Merck

Skip To

A3179

Differentiation Solution

Synonym(s):

Acid Alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 L
Check Cart for Availability
€ 73,40

About This Item

UNSPSC Code:
12171500
NACRES:
NA.47
MDL number:

€ 73,40


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

40 mo.

color

colorless

application(s)

hematology
histology

storage temp.

room temp

Application

Acidified alcohol solution for the differentiation of regressive hematoxylin stains.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A3429NIST2897ASTS0007
color

colorless

color

colorless

color

-

color

-

form

solution

form

solution

form

-

form

liquid

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

environmental

application(s)

-

shelf life

40 mo.

shelf life

40 mo.

shelf life

-

shelf life

-

storage temp.

room temp

storage temp.

room temp

storage temp.

-

storage temp.

-

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100


Still not finding the right product?

Explore all of our products under Differentiation Solution


signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Met. Corr. 1 - STOT SE 2

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

77.0 °F

flash_point_c

25 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

7647-01-0 + 64-17-5 + 67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Radoslaw Junka et al.
Materials science & engineering. C, Materials for biological applications, 113, 110981-110981 (2020-06-04)
Bone tissue engineering aims to alleviate the shortage of available autograft material and the biological/mechanical incompatibility of allografts through fabrication of bioactive synthetic bone graft substitutes. However, these substitute grafting materials have insufficient biological potency that limits their clinical efficacy



Global Trade Item Number

SKUGTIN
A3179-1L04061838203618

Questions

  1. Is this solution ready to use or has to be diluted before use?

    1 answer
    1. These products are ready-to-use. No dilution is required.

      Helpful?

Reviews

No rating value

Active Filters