Skip to Content
Merck

Skip To

A274

Monoclonal Anti-H+/K+ ATPase (β Subunit) antibody produced in mouse

clone 2G11, ascites fluid, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
100 μL
Check Cart for Availability
€ 722,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
2G11, monoclonal
Application:
IF, IHC (f), WB CL
Citations:
4

€ 722,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

2G11, monoclonal

form

buffered aqueous solution

mol wt

antigen 60-80 kDa (glycosylated form)

species reactivity

pig, mouse, rabbit, rat, bovine, ferret, canine

technique(s)

immunohistochemistry (frozen sections): suitable, indirect immunofluorescence: 1:2,000, western blot (chemiluminescent): 1:4,000

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Atp4b(11945)
rat ... Atp4b(24217)

General description

The H+/K+-ATPase is a gastric proton pump that is a member of the P-type cation-transporting adenosine 5′-triphosphatase (ATPase) family. H+/K+-ATPase consists of a large transmembrane, catalytic α subunit, that contains sites for ATP binding and phosphorylation. This ATPase also has a smaller glycoprotein (β) subunit which may play a role in maintaining the structural and functional integrity of the complex. Mouse monoclonal anti-H+/K+ ATPase (β subunit) binds to H+/K+-ATPase β subunit in cow, dog, pig, rabbit, mouse, ferret and rat tissues. By immunoblotting, the antibody detects various forms of the β subunit, including a 60-80kDa glycosylated protein, a 52kDa β subunit precursor and the 34kDa core peptide.
The antibody reacts with various forms of the H+/K+ ATPase β subunit. It binds an epitope within amino acids 1-13 or 15-28 located on the cytoplasmic side of the β subunit. The antibody has been shown to inhibit the enzymatic activity of the H+/K+-ATPase and to alter the affinity of the cytoplasmic K+ binding site. It may be used to localize and detect H+/K+ ATPase β subunit.

Immunogen

core 34 kDa H+/K+ ATPase β subunit from deglycosylated pig gastric microsomes.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)
Mouse monoclonal anti-H+/K+ ATPase (β subunit) antibody can be used for indirect immunofluorescence (1:2,000), chemiluminescent western blot (1:4,000), and immunohistochemical applications.

Physical form

Solution in phosphate buffered saline containing 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A276A275A273
species reactivity

pig, mouse, rabbit, rat, bovine, ferret, canine

species reactivity

canine, chicken, human, sheep, pig, bovine, mouse

species reactivity

rat, human, canine, avian

species reactivity

human, rat, monkey, sheep, rabbit, guinea pig, canine

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

clone

2G11, monoclonal

clone

M7-PB-E9, monoclonal

clone

9A-5, monoclonal

clone

XVIF9-G10, monoclonal

Gene Information

mouse ... Atp4b(11945)
rat ... Atp4b(24217)

Gene Information

human ... ATP1A1(476)

Gene Information

human ... ATP1A1(476)
rat ... Atp1a1(24211)

Gene Information

human ... ATP1A3(478)
rat ... Atp1a3(24213)

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

ascites fluid

antibody form

ascites fluid

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Penghong Song et al.
The Journal of biological chemistry, 286(16), 14120-14128 (2011-03-04)
Kir4.1 channels were found to colocalize with the H(+)/K(+)-ATPase throughout the parietal cell (PC) acid secretory cycle. This study was undertaken to explore their functional role. Acid secretory rates, electrophysiological parameters, PC ultrastructure, and gene and protein expression were determined
Marco Tozzi et al.
Cancers, 12(3) (2020-03-14)
Pancreatic duct cells are equipped with acid/base transporters important for exocrine secretion. Pancreatic ductal adenocarcinoma (PDAC) cells may utilize such transporters to acidify extracellular tumor microenvironment, creating a niche favoring cell proliferation, fibrosis and resistance to chemotherapy-all contributing to the
Lin Ding et al.
Frontiers in immunology, 14, 1139391-1139391 (2023-06-19)
MDSCs express SCHLAFEN 4 (SLFN4) in Helicobacter-infected stomachs coincident with spasmolytic polypeptide-expressing metaplasia (SPEM), a precursor of gastric cancer. We aimed to characterize SLFN4+ cell identity and the role of Slfn4 in these cells. Single-cell RNA sequencing was performed on



Global Trade Item Number

SKUGTIN
A274-100UL04061837811906

Questions

Reviews

No rating value

Active Filters