Skip to Content
Merck

Skip To

A1640

Adenosine 5′-O-thiomonophosphate dilithium salt

≥98%

Synonym(s):

5′-AMPS

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C10H12Li2N5O6PS
CAS Number:
Molecular Weight:
375.15
NACRES:
NA.51
PubChem Substance ID:
UNSPSC Code:
41106305
MDL number:
Assay:
≥98%
Form:
powder
Storage temp.:
−20°C


assay

≥98%

form

powder

storage temp.

−20°C

SMILES string

[Li+].[Li]SP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)ncnc23

InChI

1S/C10H14N5O6PS.2Li/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(21-10)1-20-22(18,19)23;;/h2-4,6-7,10,16-17H,1H2,(H2,11,12,13)(H2,18,19,23);;/q;2*+1/p-2/t4-,6-,7-,10-;;/m1../s1

InChI key

FZXPFDFAEUBVHA-IDIVVRGQSA-L

Application

Adenosine 5′′-O-thiomonophosphate (5′-AMPS) is a 5′-O-thiophosphate which along with adenosine 5′-O-(2-thiodiphosphate) and adenosine 5′-O-(3-thiotriphosphate) can act as inhibitors of phosphohydrolases. AMPS is a substrate for smooth muscle endothelial ecto-5′-nucleotidase. 5′APMS is a stimulator of 5-lipoxygenase activity of rat polymorphonuclear (PMN) leukocytes. Histidine triad nucleotide-binding protein 1 (HINT-1) phosphoramidase transforms nucleoside 5′-O-phosphorothioates such as 5′-AMPS to nucleoside 5′-O-phosphates.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A6779A9396A9272
form

powder

form

powder

form

powder

form

powder

assay

≥98%

assay

≥97%

assay

≥98% (HPLC)

assay

≥98% (HPLC)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 2

target_organs

Eyes,Central nervous system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
A1640-25MG04061833001875

Questions

Reviews

No rating value

Active Filters