Skip to Content
Merck

Skip To

A138

Aminobenztropine

solid

Synonym(s):

3-(2′-Aminobenzhydryloxy)tropane, ABT

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H26N2O
CAS Number:
Molecular Weight:
322.44
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Form:
solid
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

Quality Level

color

white

solubility

0.1 M HCl: soluble, 0.1 M NaOH: insoluble, DMSO: soluble, methanol: soluble

SMILES string

CN1C2CCC1CC(C2)OC(c3ccccc3)c4ccccc4N

InChI

1S/C21H26N2O/c1-23-16-11-12-17(23)14-18(13-16)24-21(15-7-3-2-4-8-15)19-9-5-6-10-20(19)22/h2-10,16-18,21H,11-14,22H2,1H3

InChI key

KZFDKINRISJFCO-UHFFFAOYSA-N

Biochem/physiol Actions

High affinity muscarinic ligand that has been used for affinity purification of muscarinic cholinergic receptors.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A143T4182D122
form

solid

form

powder

form

solid

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

solubility

0.1 M HCl: soluble, DMSO: soluble, methanol: soluble, 0.1 M NaOH: insoluble

solubility

0.1 M HCl: soluble

solubility

0.1 M HCl: soluble <0.4 mg/mL, DMSO:methanol (1:1): soluble 10 mg/mL, clear, colorless, ethanol: soluble 10 mg/mL, methylene chloride: soluble 10 mg/mL, 0.1 M NaOH: insoluble, 2-hydroxypropyl-β-cyclodextrin: insoluble, H2O: insoluble

solubility

DMSO: >10 mg/mL, 0.1 M HCl: soluble, 0.1 M NaOH: slightly soluble, H2O: insoluble

color

white

color

white to beige

color

-

color

white

Gene Information

human ... CHRM1(1128), CHRM2(1129), CHRM3(1131), CHRM4(1132), CHRM5(1133)

Gene Information

human ... ADRB1(153)

Gene Information

human ... EGFR(1956)
mouse ... Egfr(13649)

Gene Information

human ... DRD1(1812), DRD2(1813), DRD3(1814), DRD4(1815), DRD5(1816), KCNH2(3757)
rat ... Drd1a(24316), Drd2(24318), Drd3(29238), Htr3a(79246)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Victor Sebastian-Perez et al.
Journal of enzyme inhibition and medicinal chemistry, 35(1), 511-523 (2020-01-16)
A previous phenotypic screening campaign led to the identification of a quinazoline derivative with promising in vitro activity against Schistosoma mansoni. Follow-up studies of the antischistosomal potential of this candidate are presented here. The in vivo studies in a S. mansoni mouse
Tatiane Souza Dos Santos et al.
Frontiers in veterinary science, 7, 230-230 (2020-05-20)
An experiment was conducted to evaluate the effect of different levels of inorganic copper and zinc on growth performance, intestinal permeability, intestinal lesion scores, oocyst shedding, antioxidant properties and bone quality in broilers challenged with Eimeria spp. A total of
A Rinken et al.
Archives of biochemistry and biophysics, 301(1), 158-164 (1993-02-15)
Muscarinic acetylcholine receptors (mAChRs) can be solubilized from porcine atrial membranes in sucrose monolaurate (SM-1200) with retention of up to 80% of N-[3H]methylscopolamine ([3H]NMS) binding activity. The mAChRs solubilized by SM-1200 were relatively stable at 4 degrees C with an



Global Trade Item Number

SKUGTIN
A138-250MG04061832079103
A138-50MG04061832088563

Questions

Reviews

No rating value

Active Filters