Skip to Content
Merck

Skip To

A0779

p-Aminoclonidine hydrochloride

solid

Synonym(s):

2-(4-Amino-2,6-dichloroanilino)-2-imidazoline hydrochloride, Apraclonidine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 53,20
€ 45,22

About This Item

Empirical Formula (Hill Notation):
C9H10Cl2N4 · xHCl
Molecular Weight:
245.11 (free base basis)
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Form:
solid
Quality level:

€ 45,22

List Price€ 53,20Save 15%
Web-Only Promotion

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

Quality Level

color

white

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 1.4 mg/mL (Solutions may be stored for several days at 4 °C), boiling water: 100 mg/mL (Solutions may be stored for several days at 4 °C), 0.1 M HCl: soluble (Solutions may be stored for several days at 4 °C), methanol: soluble (Solutions may be stored for several days at 4 °C)

storage temp.

2-8°C

SMILES string

Cl[H].Nc1cc(Cl)c(NC2=NCCN2)c(Cl)c1

InChI

1S/C9H10Cl2N4.ClH/c10-6-3-5(12)4-7(11)8(6)15-9-13-1-2-14-9;/h3-4H,1-2,12H2,(H2,13,14,15);1H

InChI key

OTQYGBJVDRBCHC-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

α2-adrenoceptor agonist.

Features and Benefits

This compound is featured on the α2-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7897B016A1784
form

solid

form

solid

form

solid

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

200

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

−20°C

solubility

45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 1.4 mg/mL (Solutions may be stored for several days at 4 °C), 0.1 M HCl: soluble (Solutions may be stored for several days at 4 °C), methanol: soluble (Solutions may be stored for several days at 4 °C), boiling water: 100 mg/mL (Solutions may be stored for several days at 4 °C)

solubility

H2O: soluble, methanol: soluble

solubility

H2O: soluble (Aqueous solutions may be stored for several weeks at -20 °C.), ethanol: soluble, isopropanol: soluble

solubility

2 M NaOH: 50 mg/mL (Solutions may be stored for 1-2 days at 4°C.)

color

white

color

white

color

white

color

light yellow to yellow-brown

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

human ... ADRA1A(148), ADRA1B(147), ADRA1D(146)

Gene Information

human ... FPGS(2356)
mouse ... Fpgs(14287)


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Articles

Learn about alpha-2 adrenoceptor and its subtypes, mediated responses, and applications of agonists. Included is a list of available products and a comparison table.


J Cepelík et al.
Physiological research, 46(3), 203-208 (1997-01-01)
The effects of the selective alpha2-adrenergic agonist p-aminoclonidine, the nonselective adrenergic agonist epinephrine, the selective beta2-adrenergic agonist fenoterol and the adenosine A1 agonist R-PIA on intraocular pressure were studied in control and pertussis toxin-pretreated rabbits. Pretreatment of rabbits with pertussis
T Salminen et al.
The Journal of biological chemistry, 274(33), 23405-23413 (1999-08-07)
We have compared bacteriorhodopsin-based (alpha(2A)-AR(BR)) and rhodopsin-based (alpha(2A)-AR(R)) models of the human alpha(2A)-adrenengic receptor (alpha(2A)-AR) using both docking simulations and experimental receptor alkylation studies with chloroethylclonidine and 2-aminoethyl methanethiosulfonate hydrobromide. The results indicate that the alpha(2A)-AR(R) model provides a better
M E Alburges et al.
Brain research bulletin, 32(2), 97-102 (1993-01-01)
The localization of alpha 2-receptors was determined by quantitative autoradiography using [125I]para-iodoclonidine ([125I]PIC) and [3H]para-aminoclonidine ([3H]PAC). In cortical tissue, [125I]PIC and [3H]PAC were equipotent in their capacity to bind sites recognized by oxymetazoline (preferentially binds to the alpha 2A receptor



Global Trade Item Number

SKUGTIN
A0779-5MG04061832268903
A0779-1MG04061825980430

Questions

Reviews

No rating value

Active Filters