Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
Curacao aloe
Quality Level
form
powder
concentration
~50%
color
yellow to brown
mp
418.39 °C ((785.10 °F ))
solubility
pyridine: 50 mg/mL, clear, dark red to very dark red and red purple
storage temp.
room temp
SMILES string
OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@H]2c3cccc(O)c3C(=O)c4c(O)cc(CO)cc24
InChI
1S/C21H22O9/c22-6-8-4-10-14(21-20(29)19(28)17(26)13(7-23)30-21)9-2-1-3-11(24)15(9)18(27)16(10)12(25)5-8/h1-5,13-14,17,19-26,28-29H,6-7H2/t13-,14+,17-,19+,20-,21+/m1/s1
InChI key
AFHJQYHRLPMKHU-OSYMLPPYSA-N
Biochem/physiol Actions
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| biological source Curacao aloe | biological source Aloe barbadensis Miller leaves | biological source - | biological source - |
| solubility pyridine: 50 mg/mL, clear, dark red to very dark red and red purple | solubility methanol: 100 mg/mL, clear to very slightly hazy, light yellow to yellow | solubility - | solubility - |
| Quality Level 100 | Quality Level 200 | Quality Level - | Quality Level - |
| form powder | form powder | form - | form - |
| storage temp. room temp | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| mp 418.39 °C ((785.10 °F )) | mp - | mp - | mp - |
Still not finding the right product?
Explore all of our products under Aloin
signalword
Warning
hcodes
pcodes
Hazard Classifications
Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A0451-25G | 04061833333921 |

