Skip to Content
Merck

Skip To

A0326

(S)-AMPA

≥97%, AMPA receptor agonist, powder

Synonym(s):

(S)-α-Amino-3-hydroxy-5-methylisoxazole-4-propionic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 249,00
5 mg
Check Cart for Availability
€ 817,00

About This Item

Empirical Formula (Hill Notation):
C7H10N2O4
CAS Number:
Molecular Weight:
186.17
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥97%
Form:
powder

€ 249,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

(S)-AMPA, ≥97%

Quality Segment

assay

≥97%

form

powder

SMILES string

Cc1onc(O)c1C[C@H](N)C(O)=O

InChI

1S/C7H10N2O4/c1-3-4(6(10)9-13-3)2-5(8)7(11)12/h5H,2,8H2,1H3,(H,9,10)(H,11,12)/t5-/m0/s1

InChI key

UUDAMDVQRQNNHZ-YFKPBYRVSA-N

General description

AMPA (α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid) activates excitatory postsynaptic currents (EPSC).

Application

(S)-AMPA has also been used to activate rostromedial tegmental nucleus (RMTg), to study aversive conditioning in relation with cocaine-induced avoidance behaviors in mice.[1]
(S)-AMPA has been used to selectively eliminate the neural cochlear component of adult barn owls by destroying afferent synapses.

Biochem/physiol Actions

Active enantiomer of (RS)-AMPA zwitterion. Potent agonist at the AMPA subclass of ionotropic glutamate receptors.
Potent agonist at the AMPA subclass of ionotropic glutamate receptors; active enantiomer.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Glutamate Receptors (Ion Channel Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A9111A6816A263
form

powder

form

powder

form

solid

form

solid

assay

≥97%

assay

≥98% (HPLC)

assay

-

assay

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

-

Gene Information

human ... GRIA1(2890), GRIA2(2891), GRIA4(2893), GRIK1(2897)
mouse ... Gria1(14799)
rat ... Gria1(50592), Gria2(29627), Gria3(29628), Gria4(29629), Grik1(29559), Grik2(54257), Grik4(24406), Grin2a(24409), Grin2b(24410), Grm6(24419)

Gene Information

-

Gene Information

rat ... Gria1(50592), Gria2(29627), Gria3(29628), Gria4(29629), Grik1(29559), Grik2(54257), Grin2a(24409), Grm1(24414), Grm2(24415), Grm4(24417), Grm5(24418), Slc1a1(25550), Slc1a2(29482), Slc1a3(29483)

Gene Information

human ... GRIK1(2897)
rat ... Gria1(50592), Grik1(29559), Grin2a(24409)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
A0326-5MG04061832079080
A0326-1MG04061825980423

Questions

Reviews

No rating value

Active Filters