Skip to Content
Merck

Skip To

A0224

Anti-l/s-Afadin antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AF6, Anti-MLL-AF6, Anti-MLLT4, Anti-l-afadin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
0.2 mL
Check Cart for Availability
€ 526,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ARR, ICC, IF, IHC (f), WB
Citations:
29

€ 526,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

mol wt

antigen ~200 kDa (and 190 kDa)

species reactivity

rat, human, canine, mouse

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunocytochemistry: suitable, immunohistochemistry (frozen sections): 1:1,000 using mouse liver sections., indirect immunofluorescence: 1:250 using cultured dog (MDCK) cells and of cultured human HepG2 cells, indirect immunofluorescence: 1:500 using frozen sections of mouse liver, microarray: suitable, western blot: 1:1000 using extract of rat brain.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MLLT4(4301)
mouse ... Mllt4(17356)
rat ... Mllt4(26955)

General description

Afadin is a cell-cell adherens junction, F-actin binding multidomain protein. This adherens junction protein has two splicing variants: the ubiquitously expressed l-afadin, and the smaller s-afadin, which is abundantly expressed in neural tissues. Afadin associates with E-catenin proteins.
S-Afadin is a protein which lacks both the third proline-rich and the F-actin binding regions and is homologous to the human AF-6 (ALL-1 fusion partner from chromosome 6) gene product. Anti-l/s-Afadin is developed in rabbit using a synthetic peptide of rat afadin.

Immunogen

synthetic peptide corresponding to rat l/s afadin sequence, amino acids 1163-1179, with N-terminal cysteine added, conjugated to BSA.

Application

Anti-l/s-Afadin antibody produced in rabbit has been used in :
  • immunocytochemistry
  • immunohistochemistry
  • immunoprecipitation

Biochem/physiol Actions

Anti-l/s-Afadin specifically binds to l-afadin and s-afadin by immunoblotting (approximately 200 and 190 kDa respectively). Additional bands may appear in certain extract preparations in western blot assays. The immunizing peptide specifically inhibits the staining of the I/s-afadin bands. The antibody is specific for human, dog, rat, and mouse l/s-afadin.
s-Afadin binds to several cytoplasmic proteins such as the small guanosine triphosphatase (GTPase) Ras, the tight junction protein ZO-1 2 and the vinculin binding protein ponsin/ sorbin and SH3 domain containing 1 (SH3P12). In addition, l-afadin also binds through its PDZ several integral membrane components such as the Ca2+ -independent homophilic immunoglobulin-like nectin, the junctional adhesion molecule (JAM)4 and a subset of the EphB receptor protein-tyrosine kinases.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% BSA and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A0349S4945C3595
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

technique(s)

immunocytochemistry: suitable, indirect immunofluorescence: 1:250 using cultured dog (MDCK) cells and of cultured human HepG2 cells, microarray: suitable, immunohistochemistry (frozen sections): 1:1,000 using mouse liver sections., western blot: 1:1000 using extract of rat brain., indirect immunofluorescence: 1:500 using frozen sections of mouse liver

technique(s)

immunohistochemistry (frozen sections): 1:1,000 using mouse liver sections., indirect immunofluorescence: 1:500 using MDCK cells and cultured human HepG2 cells., microarray: suitable, western blot: 1:2,000 using extract of rat brain

technique(s)

immunohistochemistry: suitable using suitable for IHC on rat brain perfused with 4% paraformaldehyde, microarray: suitable, western blot: 0.5-1.0 μg/mL using mouse brain and rat brain extract (S1 fraction)

technique(s)

immunohistochemistry (frozen sections): 1:400 using frozen rat liver sections, western blot: 1:400 using a rat liver membrane preparation

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters