Skip to Content
Merck
All Photos(1)

Key Documents

95348

Sigma-Aldrich

Abberior® STAR 470SX, NHS ester

for long Stokes STED and 2-color STED application

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Assay

≥80.0% (degree of coupling)

form

solid

solubility

DMF: 1 mg/mL, clear

fluorescence

λex 480 nm; λem 610-630 nm in PBS, pH 7.4

storage temp.

−20°C

General description

Abberior STAR 470SX is the latest development of long-Stokes-Shift dyes for STED microscopy. The dye can be excited from 450 to 480nm. It can substitute dyes like Chromeo 494. For STED, a depletion wavelength ~750 nm is recommended. It is therefore well suited for 2-color STED imaging as implemented in the Leica TCS STED Ti:Sa microscope.

Abberior STAR 470SX is the dye of choice for long Stokes STED applications in the orange fluorescent regime. The dye is particularly designed and tested for 2-color STED microscopy in combination with our STAR 635 using a single STED wavelength. The dye is our recommendation for usage in the Leica TCS STED Ti:Sa 2-color system.

Key Features
  • Designed for STED microscopy at ~750 nm
  • Long Stokes′ shift (>130 nm) for 2-color applications
  • Tested in the Leica TCS STED Ti:Sa 2-color system

Absorption Maximum, λmax: 475 nm (MeOH),
477 nm (PBS, pH 7.4)
Extinction Coefficient, ε(λmax): 30′400 M-1cm-1 (MeOH),
22′700 M-1cm-1 (PBS, pH 7.4)
Correction Factor, CF260 = ε260/εmax: 0.69
Correction Factor, CF280 = ε280/max: 0.47
Fluorescence Maximum, λfl: 609 nm (MeOH),
627 nm (PBS, pH 7.4)
Recommended STED Wavelength, λSTED: 740 - 770 nm
Fluorescence Quantum Yield, η: 0.80 (EtOH)
Fluorescence Lifetime, τ: 3.9 (EtOH)

Application

Abberior® STAR 470SX goat anti-rabbit antibody has been used for STED (stimulated emission depletion) microscopy in Caco-2 cells.

Suitability

Designed and tested for fluorescent super-resolution microscopy

Legal Information

Chromeo is a trademark of Active Motif Chromeon GmbH
abberior is a registered trademark of Abberior GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thommie Karlsson et al.
PLoS pathogens, 8(10), e1002953-e1002953 (2012-10-17)
Quorum sensing (QS) signaling allows bacteria to control gene expression once a critical population density is achieved. The Gram-negative human pathogen Pseudomonas aeruginosa uses N-acylhomoserine lactones (AHL) as QS signals, which coordinate the production of virulence factors and biofilms. These
S W Hell et al.
Optics letters, 19(11), 780-782 (1994-06-01)
We propose a new type of scanning fluorescence microscope capable of resolving 35 nm in the far field. We overcome the diffraction resolution limit by employing stimulated emission to inhibit the fluorescence process in the outer regions of the excitation
Marcus Dyba et al.
Nature biotechnology, 21(11), 1303-1304 (2003-10-21)
We report immunofluorescence imaging with a spatial resolution well beyond the diffraction limit. An axial resolution of approximately 50 nm, corresponding to 1/16 of the irradiation wavelength of 793 nm, is achieved by stimulated emission depletion through opposing lenses. We
T A Klar et al.
Optics letters, 24(14), 954-956 (2007-12-13)
We overcame the resolution limit of scanning far-field fluorescence microscopy by disabling the fluorescence from the outer part of the focal spot. Whereas a near-UV pulse generates a diffraction-limited distribution of excited molecules, a spatially offset pulse quenches the excited
Volker Westphal et al.
Physical review letters, 94(14), 143903-143903 (2005-05-21)
Utilizing single fluorescent molecules as probes, we prove the ability of a far-field microscope to attain spatial resolution down to 16 nm in the focal plane, corresponding to about 1/50 of the employed wavelength. The optical bandwidth expansion by nearly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service