Skip to Content
Merck
All Photos(1)

Key Documents

8.14371

Sigma-Aldrich

Triethylamine trishydrofluoride

for synthesis

Synonym(s):

Triethylamine trishydrofluoride

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
€25.40
10 ML
€75.30

€25.40


Estimated to ship on15 April 2025


Request a Bulk Order

Select a Size

Change View
2 ML
€25.40
10 ML
€75.30

About This Item

Linear Formula:
(C2H5)3N · 3 HF
CAS Number:
Molecular Weight:
161.21
MDL number:
UNSPSC Code:
12352107
EC Index Number:
277-550-5
NACRES:
NA.22

€25.40


Estimated to ship on15 April 2025


Request a Bulk Order

Quality Level

form

liquid

pH

3 (20 °C in H2O, saturated aqueous solution)

bp

70 °C/19.9 hPa

transition temp

flash point 88 °C

density

1.00 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[F-].[F-].[F-].[N+H](CC)(CC)CC.[H+].[H+]

InChI

1S/C6H15N.3FH/c1-4-7(5-2)6-3;;;/h4-6H2,1-3H3;3*1H

InChI key

IKGLACJFEHSFNN-UHFFFAOYSA-N

Application

  • Asymmetric synthesis of fluoroamines from chiral aziridines: This study explores the use of triethylamine trishydrofluoride in the synthesis of fluoroamines, highlighting its role in enhancing reaction selectivity and efficiency (Park et al., 2014).
  • Recent advance in the synthesis of (1, 1-difluoroethyl) arenes: This paper discusses recent advancements in the synthesis of difluoroethyl arenes using triethylamine trishydrofluoride, contributing to the development of new fluorination techniques (Li et al., 2018).
  • Quantification of hydrogen fluoride-based reagents using a bifunctional NMR internal standard: Describes the quantification of reagents like triethylamine trishydrofluoride in chemical syntheses, emphasizing the accuracy of measurement techniques (Hammond et al., 2016).
  • Extensive CRISPR RNA modification reveals chemical compatibility and structure-activity relationships for Cas9 biochemical activity: This research utilizes triethylamine trishydrofluoride for RNA modifications in CRISPR-Cas9 processes, showing its potential in genetic engineering (O′Reilly et al., 2019).

Analysis Note

Assay (HClO₄): ≥ 98.0 %
Colour (visual): colourless, almost colourless
Density (d 20 °C/ 4 °C): 0.995 - 1.000
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F

Flash Point(C)

88 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service