Skip to Content
Merck
All Photos(1)

Key Documents

GE27-0846-01

Thrombin Protease

Cytiva 27-0846-01

Synonym(s):

Proteolytic Enzyme, Thrombin Activity, Thrombin Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.56

packaging

pkg of 500 units

manufacturer/tradename

Cytiva 27-0846-01

shipped in

dry ice

storage temp.

−20°C

General description

Thrombin is a protease used to digest fusion proteins prepared from pGEX vectors recognition sequence for thrombin (pGEX-1lT, pGEX-2T, pGEX-2TK, pGEX-4T-1, pGEX-4T-2, and pGEX-4T-3). Purified from bovine plasma.

Thrombin is a protease used to digest fusion proteins prepared from pGEX vectors (see pGEX Vectors (GST Gene Fusion System)) containing the recognition sequence for thrombin (pGEX-1lT, pGEX-2T, pGEX-2TK, pGEX-4T-1, pGEX-4T-2, and pGEX-4T-3).

Application

Used for site-specific separation of the GST tag from proteins expressed using pGEX-T vectors

Features and Benefits

  • Enables the site-specific cleavage of fusion proteins containing an accessible thrombin recognition sequence
  • Purified from bovine plasma; functionally free of other clotting factors, plasminogen, and plasmin
  • Can be used either following affinity purification or while fusion proteins are bound to Glutathione Sepharose 4B, Glutathione Sepharose High Performance, or Glutathione Sepharose 4 Fast Flow

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas E Wood et al.
Cell reports, 29(1), 187-201 (2019-10-03)
The type VI secretion system (T6SS) is crucial in interbacterial competition and is a virulence determinant of many Gram-negative bacteria. Several T6SS effectors are covalently fused to secreted T6SS structural components such as the VgrG spike for delivery into target
Guanglin Xing et al.
Neuron, 109(12), 1963-1978 (2021-05-26)
Our daily life depends on muscle contraction, a process that is controlled by the neuromuscular junction (NMJ). However, the mechanisms of NMJ assembly remain unclear. Here we show that Rapsn, a protein critical for NMJ formation, undergoes liquid-liquid phase separation
Moonsup Lee et al.
The Journal of cell biology, 221(1) (2021-11-18)
Proper cilia formation in multiciliated cells (MCCs) is necessary for appropriate embryonic development and homeostasis. Multicilia share many structural characteristics with monocilia and primary cilia, but there are still significant gaps in our understanding of the regulation of multiciliogenesis. Using
Sarah van Veen et al.
Nature, 578(7795), 419-424 (2020-01-31)
ATP13A2 (PARK9) is a late endolysosomal transporter that is genetically implicated in a spectrum of neurodegenerative disorders, including Kufor-Rakeb syndrome-a parkinsonism with dementia1-and early-onset Parkinson's disease2. ATP13A2 offers protection against genetic and environmental risk factors of Parkinson's disease, whereas loss

Articles

This page describes troubleshooting strategies for cloning the gene or gene fragment into a pGEX expression vector.

This page shows troubleshooting strategies for cleavage methods using Cytiva products.

This page shows how to cleave and purify GST-tagged proteins bound to Glutathione Sepharose in batch mode from Cytiva. Glutathione Sepharose High Performance, Glutathione Sepharose 4 Fast Flow, and Glutathione Sepharose 4B can all be used for cleavage and purification of GST-tagged proteins in batch.

This page shows how to cleave and purify GST-tagged proteins eluted from GSTrap from Cytiva.

See All

Protocols

This page describes the characteristics of pGEX expression vectors used with Cytiva products.

Protect fusion proteins from proteolytic degradation with enzyme removal pre- and post-cleavage using PreScission Protease, thrombin, or Factor Xa.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service