Skip to Content
Merck
All Photos(1)

Key Documents

93859

Sigma-Aldrich

BioStab immunoassay stabilizer

Synonym(s):

Immunoassay stabilizer BioStab

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

form

liquid

impurities

DNases, none detected
RNases, none detected
proteases, none detected

storage temp.

2-8°C

General description

BioStab immunoassay stabilizer is a 4x concentrate in sterile, double-distilled water.

Application

Immunoassays as (ELISA Tests) may be stabilized with BioStab immunoassay stabilizer to increase stability and improve coating of antibodies.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Open-sandwich enzyme immunoassay for one-step noncompetitive detection of corticosteroid 11-deoxycortisol.
Ihara M, Suzuki T, Kobayashi N, et al.
Analytical Chemistry, 81, 8298-8304 (2010)
Satoshi Sekiguchi et al.
Bioscience, biotechnology, and biochemistry, 76(1), 95-100 (2012-01-11)
Bovine intestine alkaline phosphatase (BIALP) is widely used as a signaling enzyme in sensitive assays such as enzyme immunoassay. In this study, we evaluated the effects of sugars on the kinetic stability of BIALP in the hydrolysis of p-nitrophenylphosphate (pNPP).
Kazuki Nishizawa et al.
Colloids and surfaces. B, Biointerfaces, 77(2), 263-269 (2010-03-04)
A phospholipid polymer platform and an antibody as a bioaffinity ligand were used to construct a biointerface for a highly sensitive immunoassay. The platform had a nanometer-scaled particle deposition surface and it was constructed with poly[2-methacryloyloxyethyl phosphorylcholine (MPC)-co-n-butyl methacrylate (BMA)-co-p-nitrophenyloxycarbonyl
A P Pirozhkov et al.
Voprosy virusologii, 55(1), 45-48 (2010-04-07)
The time course of changes in the activity of solutions of horseradish peroxidase conjugates with immunoglobulins against Ebola and Marburg fevers was studied in the presence of different components. The series of the conjugates of ELISA kits for the detection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service