Skip to Content
Merck

Skip To

18995

BioStab Peroxidase Conjugate Stabilizer

Synonym(s):

Peroxidase Conjugate Stabilizer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
250 mL
Check Cart for Availability
€ 206,00
1 L
Check Cart for Availability
€ 639,00

About This Item

NACRES:
NA.25
UNSPSC Code:
12161706

€ 206,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

technique(s)

HPLC: suitable

impurities

DNases, none detected, RNases, none detected, proteases, none detected

suitability

in accordance for identity (HPLC)

storage temp.

2-8°C

General description

BioStab Peroxidase Conjugate Stabilizer is a multi-component solution for stabilization of peroxidase conjugate at elevated temperatures and high dilutions for long periods. It does not contain any protein, thus no disturbing interactions will appear.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S107555514PNS1010
Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

200

form

liquid

form

solution

form

-

form

liquid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

suitability

in accordance for identity (HPLC)

suitability

-

suitability

corresponds for identity (HPLC)

suitability

-

impurities

DNases, none detected, proteases, none detected

impurities

-

impurities

DNases, none detected, RNases, none detected, phosphatases, none detected, proteases, none detected

impurities

-

technique(s)

HPLC: suitable

technique(s)

flow cytometry: suitable

technique(s)

HPLC: suitable

technique(s)

-


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters