Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 10 nmol | Check Cart for Availability | € 404,00 |
About This Item
NACRES:
NA.32
UNSPSC Code:
12352108
Quality Level
assay
≥90%
form
solid
mol wt
Mw 1473 g/mol
manufacturer/tradename
ATTO-TEC GmbH
fluorescence
λex 594 nm; λem 624 nm in 0.1 M phosphate pH 7.0
suitability
suitable for fluorescence
storage temp.
−20°C
General description
Atto 590 is a novel fluorescent label that belongs to the class of Rhodamine dyes. It shows a strong absorption, high fluorescence quantum yield, high thermal and photostability, and a very little triplet formation. Atto 590 consists of a mixture of two isomers with practically identical optical absorption and emission Phalloidin is a fungal toxin isolated from the poisonous mushroom Amanita phalloides. Its toxicity is attributed to the ability to bind F actin in liver and muscle cells. As a result of binding phalloidin, actin filaments become strongly stabilized. Phalloidin has been found to bind only to polymeric and oligomeric forms of actin, and not to monomeric actin. The dissociation constant of the actin-phalloidin complex has been determined to be on the order of 3 x 10-8. Phalloidin differs from amanitin in rapidity of action; at high dose levels, death of mice or rats occurs within 1 or 2 hours. Fluorescent conjugates of phalloidin are used to label actin filaments for histological applications. Some structural features of phalloidin are required for the binding to actin. However, the side chain of amino acid 7 (g-d-dihydroxyleucine) is accessible for chemical modifications without appreciable loss of affinity for actin.
find more information here
find more information here
1 of 1
This Item | |||
|---|---|---|---|
| suitability suitable for fluorescence | suitability suitable for fluorescence | suitability suitable for fluorescence | suitability suitable for fluorescence |
| fluorescence λex 594 nm; λem 624 nm in 0.1 M phosphate pH 7.0 | fluorescence - | fluorescence λex 663 nm; λem 684 nm in 0.1 M phosphate pH 7.0 | fluorescence - |
| form solid | form solid | form - | form solid |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| assay ≥90% | assay ≥80.0% (HPLC) | assay ≥90% (HPLC) | assay ≥90.0% (HPLC) |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Qiao-Chu Wang et al.
Theriogenology, 144, 132-138 (2020-01-16)
N-WASP is the mammalian ortholog of WASP which is an actin nucleation promoting factor and has been reported to regulate actin nucleation and polymerization for multiple cell activities. However, the expression and functions of N-WASP in porcine oocytes are still
Meng-Hao Pan et al.
Biology of reproduction, 102(6), 1203-1212 (2020-03-14)
Formin-like 3 (FMNL3) is a member of the formin-likes (FMNLs), which belong to the formin family. As an F-actin nucleator, FMNL3 is essential for several cellular functions, such as polarity control, invasion, and migration. However, the roles of FMNL3 during
Yu Zhang et al.
Journal of cellular physiology, 234(10), 18513-18523 (2019-03-27)
Cytoskeleton which includes microtubule and actin filaments plays important roles during mammalian oocyte maturation. In the present study, we showed that protein kinase C mu (PKC mu) was one potential key molecule which affected cytoskeleton dynamics in mouse oocytes. Our
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 93042-10NMOL | 04061837737619 |
