Skip to Content
Merck

Skip To

91783

Azo-Galactomannan from carob

galactomannanase substrate, powder

Synonym(s):

Galactomannan polysaccharide from carob seeds dyed with Remazolbrilliant Blue

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 170,00

About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

€ 170,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Azo-Galactomannan from carob, galactomannanase substrate

Quality Segment

form

powder

Application

suitable as substrate for galactomannanase

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
48230G075372762
Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

form

powder

form

powder

form

powder

form

powder


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters