Skip to Content
Merck

Skip To

91352

2,6-Difluorophenyl pentafluorophenyl adipate

≥97.0% (HPLC)

Synonym(s):

Hexanedioic acid 1-(2,6-difluorophenyl) 6-(2,3,4,5,6-pentafluorophenyl) ester

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C18H11F7O4
CAS Number:
Molecular Weight:
424.27
UNSPSC Code:
12352204
NACRES:
NA.25
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
22620419
Assay:
≥97.0% (HPLC)
Form:
lumps


assay

≥97.0% (HPLC)

form

lumps

SMILES string

O=C(OC1=C(F)C=CC=C1F)CCCCC(OC2=C(F)C(F)=C(F)C(F)=C2F)=O

InChI

1S/C18H11F7O4/c19-8-4-3-5-9(20)17(8)28-10(26)6-1-2-7-11(27)29-18-15(24)13(22)12(21)14(23)16(18)25/h3-5H,1-2,6-7H2

InChI key

FNHQYADULQQVJA-UHFFFAOYSA-N

Application

This linker enables to form heteroconjugates by linking primary amino groups from two separate biomolecules in a strictly orthogonal way. The orthogonality arises from the differential reactivity of the ester leaving groups.[1]


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Adam T Slósarczyk et al.
Journal of peptide science : an official publication of the European Peptide Society, 18(4), 261-269 (2012-03-07)
An efficient method for the heteroconjugation of biomolecules carrying free amino groups was reported previously, where mixed polyfluorophenyl diesters of dicarboxylic acids with varied aliphatic chain length were shown to be efficient reagents for the conjugation of a variety of



Global Trade Item Number

SKUGTIN
91352-5MG04061833627563
91352-50MG04061832010434
91352-25MG04061833627556
91352-10MG04061832019345

Questions

Reviews

No rating value

Active Filters