Skip to Content
Merck

Skip To

913

TRIZMAL™ 6.3 Concentrate

pH 6.3(concentrate), 1 M

Synonym(s):

Trizmal buffer, Trizma® maleate buffer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mL
Check Cart for Availability
€ 110,00

About This Item

UNSPSC Code:
12352106
NACRES:
NA.47
MDL number:

€ 110,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Suitable for use in the absence of visible microbial growth.

IVD

for in vitro diagnostic use

concentration

1 M

pH

6.3(concentrate)

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Application

For use in the Naphthol AS-D Chloroacetate Esterase Kit No. 91-C.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany
Trizmal is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
903C9331390360
concentration

1 M

concentration

0.2 M

concentration

1.0 M tris-HCl

concentration

~1.0 M in H2O

Quality Level

500

Quality Level

500

Quality Level

100

Quality Level

200

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

-

application(s)

diagnostic assay manufacturing

form

solution

form

solution

form

liquid

form

liquid

shelf life

Suitable for use in the absence of visible microbial growth.

shelf life

Suitable for use in the absence of visible microbial growth.

shelf life

-

shelf life

-

storage temp.

2-8°C

storage temp.

room temp

storage temp.

-

storage temp.

2-8°C


pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters