Skip to Content
Merck

Skip To

87787

6-(4-Methoxyphenyl)-2-methyl-3,7-dihydroimidazo[1,2-a]pyrazin-3(7H)-one hydrochloride

suitable for chemiluminescence, ≥98.0% (HPLC)

Synonym(s):

2-Methyl-6-(4-methoxyphenyl)-3,7-dihydroimidazo[1,2-a]pyrazin-3(7H)-one hydrochloride, Cypridina Luciferin methoxy-analogue, MCLA

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C14H13N3O2 · HCl
CAS Number:
Molecular Weight:
291.73
UNSPSC Code:
12352200
NACRES:
NA.32
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
5838857


description

luminescence test: Emmax465 nm, 0.1 M Tris pH 8.5; after addition of H2O2 (Chemiluminescence: in 0.1 M Tris pH 8.5; after addition of H2O2)

assay

≥98.0% (HPLC)

form

powder

solubility

DMSO: soluble, H2O: soluble

suitability

suitable for chemiluminescence

SMILES string

Cl[H].COc1ccc(cc1)C2=CN3C(=O)C(C)=NC3=CN2

InChI

1S/C14H13N3O2.ClH/c1-9-14(18)17-8-12(15-7-13(17)16-9)10-3-5-11(19-2)6-4-10;/h3-8,15H,1-2H3;1H

InChI key

KGMPQNFFCUDXSO-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Generation of chemiluminescence by reaction with super oxide O2 or singlet oxygen. Used in research of leucocyte function[1][2][3].


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Hagop Kojanian et al.
American journal of therapeutics, 23(6), e1456-e1463 (2014-05-08)
Statins have favorable effects on endothelial function partly because of their capacity to reduce oxidative stress. However, antioxidant vitamins, unlike statins, are not as cardioprotective, and this paradox has been explained by failure of vitamin antioxidants to ameliorate endoplasmic reticulum
Taeko Mizutani et al.
Journal of oleo science, 65(12), 993-1001 (2016-11-11)
Sodium lauryl sulfate (SLS), a representative anionic surfactant, is well-known to induce rough skin following single or multiple topical applications. The mechanism by which SLS induces rough skin is thought to result from the disruption of skin moisture function consisting
Keiko Suzuki et al.
Antioxidants (Basel, Switzerland), 9(6) (2020-06-13)
Bisphosphonates (BPs) are classified into two groups, according to their side chain structures, as nitrogen-containing BPs (NBPs) and non-nitrogen-containing BPs (non-NBPs). In this study, we examined the effects of NBPs and non-NBPs on inflammatory responses, by quantifying the inflammatory mediators



Global Trade Item Number

SKUGTIN
87787-10MG-F04061833278383

Questions

Reviews

No rating value

Active Filters