Skip to Content
Merck

Skip To

862

FRV-Alkaline Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 mL
Check Cart for Availability
€ 197,00

About This Item

NACRES:
NA.47
UNSPSC Code:
12352106

€ 197,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

concentration

5 mg/mL

application(s)

hematology
histology

shipped in

wet ice

storage temp.

2-8°C

Application

For use with Sigma kit 86R, for the demonstration of alkaline phosphatase in granulocytes. The procedure is often referred to as the LAP procedure.

Other Notes

Fast red violet LB base, 5 mg/mL, in hydrochloric acid, 0.4 mol/L, with stabilizer.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8613951916
concentration

5 mg/mL

concentration

4 mg/mL

concentration

1 g/dL in deionized water

concentration

12.5 mg/mL

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

form

solution

form

solution

form

solution

form

solution

shelf life

Expiry date on the label.

shelf life

Expiry date on the label.

shelf life

expiry date on label

shelf life

Expiry date on the label.

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

Quality Level

500

Quality Level

500

Quality Level

500

Quality Level

500


pictograms

CorrosionExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Met. Corr. 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



W A Mohler et al.
Proceedings of the National Academy of Sciences of the United States of America, 93(22), 12423-12427 (1996-10-29)
Complementing reporter genes provide biological indicators of coincident expression of proteins in cells. We have adapted intracistronic complementation of the Escherichia coli lacZ gene for use in mammalian cells. Enzymatic activity detectable by quantitative biochemical assay, flow cytometry, or microscopy
Nathalia Dos Santos Teixeira et al.
Animal reproduction science, 225, 106669-106669 (2020-12-29)
Anesthesia is a common practice used in fish research and aquaculture. It is important to understand anesthetic effects on the animal and tissues of interest to ensure validity of data and to improve animal welfare in research and fish production



Global Trade Item Number

SKUGTIN
157139-100G04061838346254
157139-25G04061838743176
862-10ML04061838203496
NIST86204061834187080
PL862-CGA-1EA04061832876375
PL862-CG-1EA04061832876337

Questions

Reviews

No rating value

Active Filters