Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
200 mL | Check Cart for Availability | € 122,00 |
About This Item
form
solution
Quality Segment
concentration
30 mM
pH
~3.0
storage temp.
2-8°C
SMILES string
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
InChI
1S/C6H8O7.3Na/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;/q;3*+1/p-3
InChI key
HRXKRNGNAMMEHJ-UHFFFAOYSA-K
Application
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 200 | Quality Level 100 | Quality Level 200 | Quality Level 500 |
| form solution | form liquid | form liquid | form solution |
| concentration 30 mM | concentration 1 M in H2O | concentration 4 % (w/v) | concentration 1.5 M |
| pH ~3.0 | pH 7.0-8.0 | pH - | pH 2.9-3.1 (25 °C) |
| storage temp. 2-8°C | storage temp. - | storage temp. 2-8°C | storage temp. room temp |
Still not finding the right product?
Explore all of our products under Citrate Solution
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
67-66-3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

