Skip to Content
Merck

Skip To

83196

Sodium acetate pH 4.6; 0.08 M – Ammonium sulfate 1.6 M – Glycerol 20% (v/v) solution

Synonym(s):

Crystallization solution 47/Kit No 75403

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.25
UNSPSC Code:
12352202


form

liquid

Quality Segment

pH

4.5±0.2 ( in neat)

density

1.17 g/mL at 20 °C

storage temp.

2-8°C


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
96864
form

liquid

form

liquid

density

1.17 g/mL at 20 °C

density

1.13 g/mL at 20 °C

pH

4.5±0.2 ( in neat)

pH

7.2±0.2 ( in neat)

storage temp.

2-8°C

storage temp.

2-8°C


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

7783-20-2

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters