Skip to Content
Merck

Skip To

80849

1-(4-Bromoacetamidobenzyl)ethylenediamine-N,N,N′,N′-tetraacetic acid

≥80% (HPLC)

Synonym(s):

1-(p-Bromoacetamidobenzyl)-EDTA, N,N′-{1-{{4-[(Bromoacetyl)amino]phenyl}methyl}-1,2-ethanediyl}bis[N-(carboxymethyl)glycine]

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C19H24BrN3O9
CAS Number:
Molecular Weight:
518.31
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:


Quality Segment

assay

≥80% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

OC(CN(CC(O)=O)C(CN(CC(O)=O)CC(O)=O)CC1=CC=C(NC(CBr)=O)C=C1)=O

InChI

1S/C19H24BrN3O9/c20-6-15(24)21-13-3-1-12(2-4-13)5-14(23(10-18(29)30)11-19(31)32)7-22(8-16(25)26)9-17(27)28/h1-4,14H,5-11H2,(H,21,24)(H,25,26)(H,27,28)(H,29,30)(H,31,32)

InChI key

VOQPQBGCWBEYEV-UHFFFAOYSA-N

General description

a chelate labeling reagent that conjugates with sulfhydryl groups


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters