Skip to Content
Merck

Skip To

79847

Hydroxynitrile Lyase, Arabidopsis thaliana, recombinant from E. coli

≥500 U/mL

Synonym(s):

Hydroxymandelonitrile Lyase, Arabidopsis thaliana, recombinant from E. coli

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
MDL number:
Specific activity:
≥500 U/mL
Recombinant:
expressed in E. coli


recombinant

expressed in E. coli

form

liquid

specific activity

≥500 U/mL

storage temp.

−20°C

Application

Hydroxynitrile lyases (Oxynitrilases) are used for various biotransformation processes and to convert a broad range of substrates. They may be used with bacterial nitrilases to generate biocatalysts [1].

Biochem/physiol Actions

Hydroxynitrile lyases (Oxynitrilases) catalyze the addition of HCN to aldehydes and ketones thereby forming α-hydroxynitriles, also known as cyanohydrins [1]. Product 79847, Hydroxynitrile Lyase from Arabidopsis thaliana , is used to hydrolyze mandelonitrile.

Other Notes

1 U corresponds to the amount of enzyme which converts 1 μmol mandelonitrile per minte at pH 5.0 and 25°C.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



White et al.
Plant physiology, 116(4), 1219-1225 (1998-04-29)
In the cyanogenic crop cassava (Manihot esculenta, Crantz), the final step in cyanide production is the conversion of acetone cyanohydrin, the deglycosylation product of linamarin, to cyanide plus acetone. This process occurs spontaneously at pH greater than 5. 0 or
Screening for hydroxynitrile lyases in plants
Hickel, A., Heinrich, G., et al.
Biotechnol. Tech., 11, 55-58 (1997)
Kathrin Emmi Scholz et al.
Applied and environmental microbiology, 78(14), 5025-5027 (2012-05-01)
Synthesis of chiral cyanohydrins is performed in a monophasic micro-aqueous reaction system using whole recombinant Escherichia coli cells expressing the Arabidopsis thaliana hydroxynitrile lyase (AtHNL). Microscopy studies employing a fusion of AtHNL with a flavin-based fluorescent protein (FbFP) reveal that



Global Trade Item Number

SKUGTIN
79847-1ML04061838089601

Questions

Reviews

No rating value

Active Filters