Skip to Content
Merck

Skip To

76728

Sodium acetate pH 4.6; 0.1M – Sodium formate 2M solution

Synonym(s):

Crystallization solution 34/Kit-No 82009

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.25
UNSPSC Code:
12352302


form

liquid

pH

4.6±0.2 ( in neat)

density

1.08 g/mL at 20 °C

storage temp.

2-8°C



Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters