Skip to Content
Merck

Skip To

73430

Luviquat Mono LS

~30% (w/w) solids basis

Synonym(s):

(Lauryl/myristyl)trimethylammonium methosulfate, Cocotrimonium Methosulfate

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12161900


description

cationic

form

liquid

concentration

~30 % (w/w) (solids)

Legal Information

Luviquat is a trademark of BASF SE


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

>212.0 °F

flash_point_c

> 100 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters