Skip to Content
Merck

Skip To

69037

Ureidosuccinic acid

98.0-102.0% (T)

Synonym(s):

N-Carbamoyl-DL-aspartic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
€ 692,00
500 mg
Check Cart for Availability
€ 1.260,00

About This Item

Linear Formula:
HOOCCH2CH(NHCONH2)COOH
CAS Number:
Molecular Weight:
176.13
UNSPSC Code:
12352106
NACRES:
NA.25
PubChem Substance ID:
EC Number:
213-096-6
Beilstein/REAXYS Number:
1726861
MDL number:

€ 692,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

98.0-102.0% (T)

mp

~175 °C (dec.)

format

neat

SMILES string

NC(=O)NC(CC(O)=O)C(O)=O

InChI

1S/C5H8N2O5/c6-5(12)7-2(4(10)11)1-3(8)9/h2H,1H2,(H,8,9)(H,10,11)(H3,6,7,12)

InChI key

HLKXYZVTANABHZ-UHFFFAOYSA-N

Biochem/physiol Actions

Metabolite of pyrimidine biosynthesis, alanine, aspartate and glutamate metabolism.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
9429550170L6661
format

neat

format

-

format

-

format

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

200

assay

98.0-102.0% (T)

assay

≥98.0% (T)

assay

≥99.0%

assay

88.0-92.0%

mp

~175 °C (dec.)

mp

170-175 °C (dec.)

mp

195-210 °C (dec.)

mp

-


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters