Skip to Content
Merck

Skip To

55819

Atto 680-Biotin

BioReagent, suitable for fluorescence, ≥90.0% (HPCE)

Synonym(s):

Biotin-Atto 680

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32
MDL number:


product line

BioReagent

Quality Segment

assay

≥90.0% (HPCE)

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 680 nm; λem 695 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

InChI

1S/C42H57N7O7S2/c1-4-48-18-10-12-27-20-30-35(22-33(27)48)56-36-23-34-29(21-31(36)45-30)28(26-58(53,54)55)24-42(2,3)49(34)19-11-15-39(51)44-17-9-5-8-16-43-38(50)14-7-6-13-37-40-32(25-57-37)46-41(52)47-40/h20-24,32,37,40H,4-19,25-26H2,1-3H3,(H4-,43,44,46,47,50,51,52,53,54,55)/t32-,37?,40-/m0/s1

InChI key

SMZVEGAKSTTZDO-QYIPWQSGSA-N

Application

Atto 680 belongs to a new generation of fluorescent labels. The dye is designed for application in the area of life science, e.g. labeling of DNA, RNA or proteins. Characteristic features of the label are strong absorption, good fluorescence quantum yield, excellent thermal and photo-stability, and very little triplet formation. Atto 680 is a zwitterionic dye with a net electrical charge of zero. The fluorescence is efficiently quenched by electron donors like guanine, tryptophan, etc. Biotin conjugates can be used in applications like ELISA or immunohistochemistry, in situ hybridization, flow cytometry and others, to identify streptavidin, avidin, or extravidin-conjugates.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters