Skip to Content
Merck

Skip To

47255

Lumilux Green D 254

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 g
Check Cart for Availability
€ 68,70

About This Item

UNSPSC Code:
12171500
NACRES:
NA.32
MDL number:

€ 68,70


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


solubility

H2O: insoluble

Quality Level


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
125660B6756B4014
solubility

H2O: insoluble

solubility

-

solubility

H2O: 1 mg/mL

solubility

H2O: 1 mg/mL, blue

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
47255-1G-F04061826661024
47255-10G-F04061832532202

Questions

Reviews

No rating value

Active Filters