Skip to Content
Merck

Skip To

43868

4-O-[2-(Acetylamino)-2-deoxy-β-D- galactopyranosyl]-D-galactopyranose

≥95% (TLC)

Synonym(s):

β-GalNAc-(1→4)-Gal, Receptor for pili of Pseudomonas aeruginosa

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C14H25NO11
CAS Number:
Molecular Weight:
383.35
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352204
MDL number:


Quality Level

assay

≥95% (TLC)

form

powder or crystals

storage temp.

−20°C

SMILES string

CC(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CO)O[C@H]1O[C@@H]2[C@H](O)[C@@H](O)C(O)O[C@@H]2CO

InChI

1S/C14H25NO11/c1-4(18)15-7-9(20)8(19)5(2-16)25-14(7)26-12-6(3-17)24-13(23)11(22)10(12)21/h5-14,16-17,19-23H,2-3H2,1H3,(H,15,18)/t5-,6-,7-,8+,9-,10-,11-,12+,13?,14+/m1/s1

InChI key

DRHGSSXSNNHAAL-KMJSDMRTSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters