Skip to Content
Merck

Skip To

38896

5-Bromo-4-chloro-3-indolyl-myo-inositol 1-phosphate ammonium salt

≥95.0% (HPLC)

Synonym(s):

X-myo-inositol phosphate

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C14H16BrClNO9P · NH3
CAS Number:
Molecular Weight:
505.64
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83


assay

≥95.0% (HPLC)

form

powder

SMILES string

[NH4+].O[C@@H]1[C@@H](O)[C@@H](O)[C@@H](OP([O-])(=O)Oc2c[nH]c3ccc(Br)c(Cl)c23)[C@H](O)[C@H]1O

InChI

1S/C14H16BrClNO9P.H3N/c15-4-1-2-5-7(8(4)16)6(3-17-5)25-27(23,24)26-14-12(21)10(19)9(18)11(20)13(14)22;/h1-3,9-14,17-22H,(H,23,24);1H3/t9?,10-,11+,12-,13-,14?;/m1./s1

InChI key

HIFQQHVVHCJEPZ-STHBLVJOSA-N

General description

Sold for research purposes only, Biosynth patents WO 98/38332, US 6068988 for the detection of microbial metabolites with a 3-indoxyl-myo-inositol-1-phosphate compound.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters