Skip to Content
Merck

Skip To

38739

Dextran sulfate sodium salt from Leuconostoc spp.

low sulfate content

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12352201
MDL number:


form

powder

quality

low sulfate content

mol wt

Mw ~10,000

composition

sulfur content, 8-13%

solubility

water: soluble

storage temp.

room temp

General description

Dextran is a polymer of anhydroglucose. It is composed of approximately 95% alpha-D-(166) linkages. The remaining (163) linkages account for the branching of dextran.

Application

Dextran sulfate is routinely used for the selective precipitation of lipoproteins, probe hybridization membrane-immobilized DNA, the release of DNA from DNA-histone complexes, and the inhibition of RNA binding to ribosomes.

Preparation Note

Sigma dextrans are derived from Leuconostoc mesenteroides.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters