Skip to Content
Merck

Skip To

30972

Abberior® STAR 635, phalloidin

for STED application

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
20 μg
Check Cart for Availability
€ 748,00

About This Item

NACRES:
NA.32
UNSPSC Code:
12352116

€ 748,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

mol wt

Mw 1165 g/mol

fluorescence

λex 634 nm; λem 654 nm±10 nm in PBS, pH 7.4

storage temp.

−20°C

Application

  • Abberior® STAR 635, phalloidin has been used for fluorescence microscopy imaging of fixed NK (natural killer) cells.[1]
  • Abberior® STAR 635 goat anti-rabbit antibody has been used for STED (stimulated emission depletion) imaging of primary cultured striatal neurons obtained from rats.[2]
  • Abberior® STAR 635 anti-rabbit antibody has been used for STED imaging of nonsensory supporting cells obtained from cochlea of rats and mice.[3]
  • Abberior® STAR 635 conjugated with secondary antibody has been used for STED imaging of microtubules in cells.[4]
Designed and tested for fluorescent super-resolution microscopy

Legal Information

abberior is a registered trademark of Abberior GmbH

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
046830767904497
fluorescence

λex 634 nm; λem 654 nm±10 nm in PBS, pH 7.4

fluorescence

λex 638 nm; λem 651 nm±10 nm in H2O

fluorescence

λex 634 nm; λem 654 nm±10 nm in PBS, pH 7.4

fluorescence

λex 663 nm; λem 684 nm in 0.1 M phosphate pH 7.0

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

mol wt

Mw 1165 g/mol

mol wt

-

mol wt

Mw 1030 g/mol

mol wt

-


pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Hans Blom et al.
PloS one, 8(9), e75155-e75155 (2013-09-24)
The phosphoprotein DARPP-32 (dopamine and cyclic adenosine 3´, 5´-monophosphate-regulated phosphoprotein, 32 kDa) is an important component in the molecular regulation of postsynaptic signaling in neostriatum. Despite the importance of this phosphoprotein, there is as yet little known about the nanoscale
Jenu Varghese Chacko et al.
Journal of biomedical optics, 19(10), 105003-105003 (2014-10-08)
Atomic force microscopes (AFM) provide topographical and mechanical information of the sample with very good axial resolution, but are limited in terms of chemical specificity and operation time-scale. An optical microscope coupled to an AFM can recognize and target an
Arnaud P Giese et al.
Development (Cambridge, England), 139(20), 3775-3785 (2012-09-20)
Vangl2 is one of the central proteins controlling the establishment of planar cell polarity in multiple tissues of different species. Previous studies suggest that the localization of the Vangl2 protein to specific intracellular microdomains is crucial for its function. However



Global Trade Item Number

SKUGTIN
30972-20UG04061833646298

Questions

Reviews

No rating value

Active Filters