Skip to Content
Merck

Skip To

3015

Transcreener® AMP2/GMP2 FP Assay

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Check Cart for Availability
€ 819,00
1 ea
Check Cart for Availability
€ 4.920,00

About This Item

NACRES:
NA.84
UNSPSC Code:
41122100

€ 819,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shipped in

dry ice

storage temp.

−20°C

General description

The Transcreener® AMP2/GMP2 FP Assay is a universal biochemical HTS assay for enzymes that produce AMP or GMP, including ubiquitin, SUMO, nucleic acid and protein ligases, phosphodiesterases (PDEs), and synthetases. Enzyme activity is signaled by a decrease in fluorescence polarization as the bound tracer is displaced from the Transcreener® AMP2/GMP2 Antibody. The assay is a simple single step mix-and-read format enabling the use of unmodified native substrate concentrations of 1 – 1000 μM. The assay provides excellent signal at low substrate conversion, with a Z′ =0.7 and =100 millipolarization shift (mP) under normal reaction conditions.

View full Transcreener® product list

Physical form

Kit with buffered aqueous solutions

Preparation Note

3015-1K = 1,000 assay, 384-well
3015-10K = 10,000 assay, 384-well

Legal Information

Transcreener is a registered trademark of BellBrook Labs

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
301830133022
shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters