Skip to Content
Merck

Skip To

29396

6-Cyclohexylhexyl β-D-maltoside

≥99.0% (TLC)

Synonym(s):

CYMAL®-6

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1 g
Check Cart for Availability
€ 387,00

About This Item

Empirical Formula (Hill Notation):
C24H44O11
CAS Number:
Molecular Weight:
508.60
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161900
MDL number:

€ 387,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

non-ionic

assay

≥99.0% (TLC)

form

solid

optical activity

[α]20/D +47.0±3°, c = 0.5% in methanol

mol wt

micellar average mol wt 32,000

aggregation number

63

technique(s)

protein purification: suitable, protein quantification: suitable

CMC

0.56 mM

SMILES string

OC[C@H]1O[C@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](OCCCCCCC3CCCCC3)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C24H44O11/c25-12-15-17(27)18(28)20(30)24(33-15)35-22-16(13-26)34-23(21(31)19(22)29)32-11-7-2-1-4-8-14-9-5-3-6-10-14/h14-31H,1-13H2/t15-,16-,17-,18+,19-,20-,21-,22-,23-,24-/m1/s1

InChI key

WUCWJXGMSXTDAV-QKMCSOCLSA-N

Application

Detergent for the purification and crystallization of membrane-bound proteins in native structure. It has a higher CMC (critical micelle concentration) and is more hydrophobic than the corresponding linear chain analog (C24).[1]

Legal Information

No license is conveyed with the purchase of this product.
CYMAL is a registered trademark of Affymetrix, Inc.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
96193D539474716
technique(s)

protein purification: suitable, protein quantification: suitable

technique(s)

protein purification: suitable, protein quantification: suitable

technique(s)

protein purification: suitable, protein quantification: suitable

technique(s)

protein quantification: suitable

form

solid

form

powder

form

powder

form

crystals

assay

≥99.0% (TLC)

assay

≥98.0% (TLC)

assay

≥98% (GC)

assay

≥98.0% (TLC)

aggregation number

63

aggregation number

66

aggregation number

-

aggregation number

-

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

100

CMC

0.56 mM

CMC

2.4-5 mM (20-25°C)

CMC

2.2

CMC

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

A guide displaying detergents' specifications and applications. The detergents are seperated into four groups (nonionic, anionic, cationic, zwitterionic) based on their hydrophillic head group. The guide also contains Non-Detergent Sulfobetaines.


C Ostermeier et al.
Current opinion in structural biology, 7(5), 697-701 (1997-11-05)
Five new membrane protein structures have been determined since 1995 using X-ray crystallography: bacterial light-harvesting complex; bacterial and mitochondrial cytochrome c oxidases; mitochondrial bc1 complex; and alpha-hemolysin. These successes are partly based on advances in the crystallization procedures for integral



Global Trade Item Number

SKUGTIN
29396-1G04061826269725

Questions

Reviews

No rating value

Active Filters