Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
NKI/C3, monoclonal
Application:
IHC (p)
Citations:
5
Pricing and availability is not currently available.
biological source
mouse
conjugate
unconjugated
antibody form
diluted ascites fluid
antibody product type
primary antibodies
clone
NKI/C3, monoclonal
description
For In Vitro Diagnostic Use in Select Regions (See Chart)
form
buffered aqueous solution
species reactivity
human
packaging
vial of 0.1 mL concentrate (263M-14), vial of 0.5 mL concentrate (263M-15), bottle of 1.0 mL predilute (263M-17), vial of 1.0 mL concentrate (263M-16), bottle of 7.0 mL predilute (263M-18)
manufacturer/tradename
Cell Marque®
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500
isotype
IgG1κ
control
melanoma
shipped in
wet ice
storage temp.
2-8°C
visualization
cytoplasmic, membranous
Gene Information
human ... CD63(967)
General description
This antibody reacts with a 53 kDa protein which forms a part of the family of tetraspan moieties. The antigen was originally designated as a lysosomal membrane protein characterized as an activation dependent platelet surface antigen. In fact the CD63 antigen has a diverse distribution on the surface and in the cytoplasm of many cell types including lymphoid, myeloid, endothelial cells and melanoma. It has been quite useful in identifying malignant melanoma. Mete et al. found it useful in differentiating renal oncocytomas (RO) from eosinophilic renal cell carcinomas (eRCC). In his series of 35 ROs, 94% demonstrated apical and/or polar CD63 positivity. Of his 77 eRCCs, 96% showed diffuse cytoplasmic staining.
Physical form
Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide
Preparation Note
Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.
Analysis Note
![]() IVD | ![]() IVD | ![]() IVD | ![]() RUO |
Other Notes
CD63 Positive Control Slides, Product No. 263S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).
For Technical Service please contact: 800-665-7284 or email: [email protected]
Legal Information
Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 263M-16 | 0841683131526 |
| 263M-17 | 0841683131533 |
| 263M-18 | 0841683131540 |
| 263M-15 | 0841683131519 |
| 263M-14 | 0841683131502 |



