Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC (p)
Citations:
8
Pricing and availability is not currently available.
biological source
rabbit
conjugate
unconjugated
antibody form
Ig fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
description
For In Vitro Diagnostic Use in Select Regions (See Chart)
form
buffered aqueous solution
species reactivity
human
packaging
vial of 0.1 mL concentrate (223A-14), vial of 0.5 mL concentrate (223A-15), bottle of 1.0 mL predilute (223A-17), vial of 1.0 mL concentrate (223A-16), bottle of 7.0 mL predilute (223A-18)
manufacturer/tradename
Cell Marque®
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:500-1:2000
control
tonsil
shipped in
wet ice
storage temp.
2-8°C
visualization
cytoplasmic
General description
Alpha-1-Antitrypsin is a protease inhibitor from the serpin superfamily, which inhibits a number of protease enzymes from inflammatory cells. It is an acute-phase plasma protein predominantly produced in hepatocytes, although it is also made in enterocytes and some mononuclear white blood cells. Alpha-1-antitrypsin immunohistochemistry is used to detect the accumulation of alpha-1-antitrypsin in hepatic lesions and histiocytic tumors.[1]
The immunohistochemical staining of Alpha-1-Antitrypsin is considered to be very useful in the study of inherited AAT deficiency, benign and malignant hepatic tumors, and yolk sac carcinomas. Positive staining for A-1-Antitrypsin may also be used in the detection of benign and malignant lesions of a histiocytic nature. The sensitivity and specificity of the results have made this antibody a useful tool in the screening of patients with cryptogenic cirrhosis or other forms of liver disease with portal fibrosis of uncertain etiology.
The immunohistochemical staining of Alpha-1-Antitrypsin is considered to be very useful in the study of inherited AAT deficiency, benign and malignant hepatic tumors, and yolk sac carcinomas. Positive staining for A-1-Antitrypsin may also be used in the detection of benign and malignant lesions of a histiocytic nature. The sensitivity and specificity of the results have made this antibody a useful tool in the screening of patients with cryptogenic cirrhosis or other forms of liver disease with portal fibrosis of uncertain etiology.
Physical form
Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide
Preparation Note
Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.
Analysis Note
![]() IVD | ![]() IVD | ![]() IVD | ![]() RUO |
Other Notes
A-1-Antitrypsin Positive Control Slides , Product No. 223S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).
For Technical Service please contact: 800-665-7284 or email: [email protected]
Legal Information
Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 223A-14 | 0841683128045 |
| 223A-17 | 0841683128076 |
| 223A-15 | 0841683128052 |
| 223A-16 | 0841683128069 |
| 223A-18 | 0841683128083 |



