Skip to Content
Merck

Skip To

21118

Calcium citrate tribasic tetrahydrate

BioUltra, ≥98.0% (calc. on dry substance, KT)

Synonym(s):

Citric acid calcium salt, Tricalcium dicitrate

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
Ca3(C6H5O7)2 · 4H2O
CAS Number:
Molecular Weight:
570.49
UNSPSC Code:
12161700
NACRES:
NA.25
PubChem Substance ID:
EC Number:
212-391-7
Beilstein/REAXYS Number:
3924520
MDL number:
Assay:
≥98.0% (calc. on dry substance, KT)
Grade:
reagent grade
Form:
powder
Solubility:
0.1 M HCl: 0.01 M at 20 °C, clear, colorless


grade

reagent grade

product line

BioUltra

assay

≥98.0% (calc. on dry substance, KT)

form

powder

impurities

Insoluble matter, passes filter test

loss

10-14% loss on drying, 180°C, 4 h

solubility

0.1 M HCl: 0.01 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤100 mg/kg

cation traces

As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤50 mg/kg, Hg: ≤0.1 mg/kg, K: ≤500 mg/kg, Li: ≤5 mg/kg, Mg: ≤1000 mg/kg, Mn: ≤50 mg/kg, Mo: ≤5 mg/kg, Na: ≤100 mg/kg, Ni: ≤5 mg/kg, Pb: ≤1 mg/kg, Sr: ≤100 mg/kg, Zn: ≤5 mg/kg

λ

0.01 M in 0.1 M HCl

UV absorption

λ: 260 nm Amax: 0.04, λ: 280 nm Amax: 0.03

SMILES string

O.O.O.O.OC1(CC(=O)O[Ca]OC(=O)C1)C(=O)O[Ca]OC(=O)C2(O)CC(=O)O[Ca]OC(=O)C2

InChI

1S/2C6H8O7.3Ca.4H2O/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;;4*1H2/q;;3*+2;;;;/p-6

InChI key

LNIZKKFWMDARJV-UHFFFAOYSA-H



Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Morton G Burt et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 28(2), 412-418 (2012-09-20)
Calcium supplements have been associated with an increased risk of cardiovascular events. However, the validity of these findings has been questioned. A major concern is that the mechanism underlying an increase in cardiovascular events has not been demonstrated. Calcium initiates
José Manuel Quesada Gómez et al.
Clinical drug investigation, 31(5), 285-298 (2011-03-17)
The combination of calcium with vitamin D (vitamin D(3) [colecalciferol]) forms the basis of preventive and therapeutic regimens for osteoporosis. A number of studies have suggested that the combination of calcium and vitamin D is effective when administered at respective
Ram Sudheer Adluri et al.
Molecular and cellular biochemistry, 340(1-2), 73-80 (2010-03-10)
Calcium is an essential mineral to support bone health and serves as a major therapeutic intervention to prevent and delay the incidence of osteoporosis. Many individuals do not obtain the optimum amount of calcium from diets and depend on bioavailable



Global Trade Item Number

SKUGTIN
21118-1KG-F04061826714515
21118-250G-F04061833440162

Questions

Reviews

No rating value

Active Filters