Skip to Content
Merck

Skip To

14136

tris-NTA Amine trifluoroacetate salt solution

≥80.0% (HPLC)

Synonym(s):

tris-NTA trifluoracetic acid salt, tris-NTA trifluoroacetate salt solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 380,00
€ 266,00

About This Item

Empirical Formula (Hill Notation):
C43H68N8O22
CAS Number:
Molecular Weight:
1049.04
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32

€ 266,00

List Price€ 380,00Save 30%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥80.0% (HPLC)

form

liquid

storage temp.

2-8°C

SMILES string

OC(CN(CC(O)=O)[C@H](C(O)=O)CCC(N1CCCN(C(CC[C@@H](C(O)=O)N(CC(O)=O)CC(O)=O)=O)CCN(C(CCCCCN)=O)CCCN(C(CC[C@@H](C(O)=O)N(CC(O)=O)CC(O)=O)=O)CC1)=O)=O

InChI

1S/C43H68N8O22/c44-13-3-1-2-6-31(52)45-14-4-15-47(33(54)11-8-29(42(70)71)50(24-37(60)61)25-38(62)63)20-21-48(34(55)12-9-30(43(72)73)51(26-39(64)65)27-40(66)67)17-5-16-46(19-18-45)32(53)10-7-28(41(68)69)49(22-35(56)57)23-36(58)59/h28-30H,1-27,44H2,(H,56,57)(H,58,59)(H,60,61)(H,62,63)(H,64,65)(H,66,67)(H,68,69)(H,70,71)(H,72,73)/t28-,29-,30-/m0/s1

InChI key

SRHQOOQNBLZULK-DTXPUJKBSA-N

Application

  • Reversible labeling of proteins or cell surfaces
  • Detection and analysis of target molecules
  • Immobilization of proteins, lipids and cells on surfaces
  • Purification and sample preparation of proteins
  • Coupling with microscopic or spectroscopic probes (e.g., fluorophores)

Features and Benefits

  • A complex of three NTA groups ensures high-affinity binding of His-tags
  • Binding affinity is approximately four orders of magnitude higher than monovalent metal ion chelators
  • Protein binding is stochiometric
  • tris-NTA is available as amino-group or biotin conjugates

Legal Information

This product was produced by Biotechrabbit GmbH. The use of this product for research purposes is covered by a license from QIAGEN Group. No rights to use this product to perform or offer diagnostic, companion diagnostic, commercial testing or other commercial services for money or money′s worth are granted by the supply of this product expressly, by implication or by estoppel.
Should you wish to use this product for any other purpose not covered by this license, please contact QIAGEN Corporate Business Development at [email protected].

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
755438.18003238732
form

liquid

form

liquid

form

powder

form

crystals

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

assay

≥80.0% (HPLC)

assay

≥80.0% (HPLC)

assay

-

assay

98%

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

-


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters