Skip to Content
Merck

Skip To

11451

N-(7-Nitrobenzofurazan-4-yl)phallacidin

suitable for fluorescence

Synonym(s):

N-(7-Nitrobenz-2-oxa-1,3-diazol-4-yl)phallacidin, NBD phallacidin

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C45H57N13O15S
CAS Number:
Molecular Weight:
1052.08
UNSPSC Code:
12171500


fluorescence

λex 464 nm; λem 536 nm in methanol

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CC(C)C1NC(=O)C(CC(C)(O)CO)NC(=O)C2Cc3c(SCC(NC(=O)C(NC1=O)C(O)C(=O)NCCNc4ccc([N+]([O-])=O)c5nonc45)C(=O)N6CC(O)CC6C(=O)NC(C)C(=O)N2)[nH]c7ccccc37

Preparation Note

Reconstitution in 1.5 mL methanol will yield a ~6.6 μM stock solution. This stock solution is sufficient for 300 applications (dying of formaldehyde-fixed cells).

Other Notes

Probe for actin: In situ localization of actin filaments; staining of F-actin


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



M.V. Parthasarathy
Plant Molecular Biology, 5, 251-251 (1987)
T. Lee et al.
Acta Histochemica Et Cytochemica, 26, 245-245 (1993)



Global Trade Item Number

SKUGTIN
157767-1L-PC04061837631931
278505-100ML04061838145635
157767-1L04061838743589
157767-25ML04061837631948