Skip to Content
Merck

Skip To

05937

N,N-Dimethylallylamine

Synonym(s):

3-Dimethylamino-1-propene, N-Allyl-dimethylamin, DMAA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH2=CHCH2N(CH3)2
CAS Number:
Molecular Weight:
85.15
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352116
EC Number:
218-458-7
MDL number:
Beilstein/REAXYS Number:
1734044
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

density

0.719 g/mL at 20 °C (lit.)

InChI

1S/C5H11N/c1-4-5-6(2)3/h4H,1,5H2,2-3H3

InChI key

GBCKRQRXNXQQPW-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
256226226289127124
density

0.719 g/mL at 20 °C (lit.)

density

0.765 g/mL at 25 °C (lit.)

density

0.732 g/mL at 25 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under N,N-Dimethylallylamine


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

-4.0 °F - closed cup

flash_point_c

-20 °C - closed cup

ppe

Faceshields, Gloves, Goggles



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



G Braunitzer et al.
Biological chemistry Hoppe-Seyler, 368(3), 195-198 (1987-03-01)
The currently used base for gas-phase sequencing, trimethylamine, can be substituted by more hydrophobic tertiary amines without any problems. We used mainly N,N-dimethylallylamine and 3-(dimethylamino)propyne, both as aqueous solutions at constant temperature (40 degrees C). The advantages of these bases
František Bílek et al.
Colloids and surfaces. B, Biointerfaces, 102, 842-848 (2012-10-30)
Three monomers (allylamine, N-allylmethylamine and N,N-dimethylallylamine) were used for grafting onto air plasma activated LDPE surface. Antibacterial agent triclosan was anchored on such substrates. Influence of graft type on the antibacterial properties was determined. Increase of antibacterial activity and amount



Global Trade Item Number

SKUGTIN
316539-25G04061833021422
05937-25ML04061838643001

Questions

Reviews

No rating value

Active Filters