Skip to Content
Merck

Skip To

05482

Bovine Serum Albumin

fraction V, ≥96% (GE)

Synonym(s):

Albumin bovine serum, BSA, Bovine albumin

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-936-2
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
≥96% (GE)
Form:
lyophilized powder


biological source

bovine

assay

≥96% (GE)

form

lyophilized powder

mol wt

Mr ~67000

purified by

heat shock fractionation

loss

≤5% loss on drying

pH

5.0-6.0 (10 mg/mL in H2O)

solubility

H2O: 40 mg/mL, clear to faintly turbid, faintly yellow to greenish-yellow, H2O: slightly soluble

storage temp.

2-8°C

Application

Albumin from bovine serum has been used as a test compound for detecting the weaker, distal chromatin co-associations by using enhanced 4C (e4C, enhanced chromosome conformation capture on chip) technique.[1]

Biochem/physiol Actions

Bovine serum albumin also referred to as BSA or Fraction V is a protein isolated from cows. BSA can block vacant binding sites in enzyme-linked immunosorbent assay (ELISA) in both poly-l-lysine (PLL)-treated as well as non-treated microwells.[2] It can also trigger insulin dependent diabetes mellitus in humans.[3]
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies.

Preparation Note

By a modified Cohn procedure, using cold ethanol, pH and low temperature precipitation.
Serum albumin may be referred to as Fraction V. This naming convention is taken from the original Cohn method of fractionating serum proteins using cold ethanol precipitation. Serum albumin was found in the fifth ethanol fraction using Cohn′s method. Since then, the term "Fraction V" has been used by some to describe serum albumin regardless of the method of preparation. Others have used this term to describe serum albumin purified by ethanol fractionation methods that have been highly modified since the original Cohn method was described. Sigma-Aldrich manufactures and distributes serum albumins purified from a variety of primary methods including the true Cohn fractionation method, modified ethanol fractionation methods, heat shock and chromatography. Additional purification steps may include crystallization or charcoal filtration.

Other Notes

As chiral complexing agent in the resolution of dansyl DL amino acids[4]
Sales restrictions may apply


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



L. Lepri et al.
Chromatographia, 36, 297-297 (1993)
J Karjalainen et al.
The New England journal of medicine, 327(5), 302-307 (1992-07-30)
Cow's milk has been implicated as a possible trigger of the autoimmune response that destroys pancreatic beta cells in genetically susceptible hosts, thus causing diabetes mellitus. Studies in animals have suggested that bovine serum albumin (BSA) is the milk protein
M Steinitz
Analytical biochemistry, 282(2), 232-238 (2000-06-30)
ELISA provides a highly sensitive procedure for quantitating antigens and antibodies. In that assay, microwells are coated initially with a specific ligand and then saturated with inert molecules to minimize nonspecific background. Coating can be improved by pretreating the microwells



Global Trade Item Number

SKUGTIN
05482-100G04061838637543
05482-25G04061838637550
05482-500G04061833438633

Questions

Reviews

No rating value

Active Filters