Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
5 mL | Check Cart for Availability | € 20,80 |
About This Item
NACRES:
NA.75
UNSPSC Code:
12352207
€ 20,80
Check Cart for Availability
form
liquid
Quality Segment
manufacturer/tradename
Cell Applications, Inc
shipped in
wet ice
storage temp.
2-8°C
Application
Thaw Neuron Coating Solution at room temperature. Pipette a sufficient amount of the Neuron Coating Solution to culture ware to cover the entire culture surface. Incubate the culture ware at 37oC for at least one hour or overnight.
Aspirate the Neuron Coating Solution from the culture ware. Rinse the culture surface twice with sterile PBS prior to use to remove unbound Neuron Coating Solution.
Aspirate the Neuron Coating Solution from the culture ware. Rinse the culture surface twice with sterile PBS prior to use to remove unbound Neuron Coating Solution.
1 of 1
This Item | |||
|---|---|---|---|
| form liquid | form liquid | form liquid | form solution |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| manufacturer/tradename Cell Applications, Inc | manufacturer/tradename Cell Applications, Inc | manufacturer/tradename - | manufacturer/tradename - |
signalword
Danger
hcodes
Hazard Classifications
Repr. 1B
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk
WGK 1
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
10043-35-3
CAS No.
10043-35-3
CAS No.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




