Skip to Content
Merck

Skip To

01971

Atto Oxa12 maleimide

BioReagent, suitable for fluorescence

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200


product line

BioReagent

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 663 nm; λem 684 nm

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto Oxa12 maleimide ester is a red-emitting dye and a lipophilic variety of Atto 655. Characteristic features of the label are strong absorption, good fluorescence quantum yield, very little triplet formation, excellent thermal and photo-stability, and outstanding ozone resistance.
Atto Oxa12 maleimide is a cationic dye (charge +1). Absorption and fluorescence are pH independent in the range pH 2 to 11, used in typical applications. Atto Oxa12 maleimide shows excellent solubility in organic solvents like DMF, DMSO or acetonitrile.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
B59802-5G04061836823559
B59802-25G04061836823542
01971-1MG04061823363563

Questions

Reviews

No rating value

Active Filters