Skip to Content
Merck

Skip To

01407

Atto 665 maleimide

BioReagent, suitable for fluorescence, ≥90% (coupling to thiols)

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200


product line

BioReagent

assay

≥90% (coupling to thiols)

mol wt

Mw 845 g/mol

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 663 nm; λem 684 nm in 0.1 M phosphate pH 7.0

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 665 is a new fluorescent label closely related to Atto 647N. It shows extraordinary high fluorescence quantum yield, excellent thermal and photo-stability, outstanding ozone resistance, and very little triplet formation. Atto 665 is a cationic (charge +1). The dye is moderately hydrophilic. The maleimide of the dye is suitable for labeling sulfhydryl (thiol) groups of proteins, in particular. Cystein residues. Prepare labeling solution of maleimide-derivative immediately before use by dissolving the vial content in anhydrous and amine-free DMF or DMSO.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Atto dyes are a series of fluorescent dyes that meet the critical needs of modern fluorescent technologies.





Global Trade Item Number

SKUGTIN
T25801-25G04061837342455
T25801-5G04061837342479
T25801-100G04061831827552

Questions

Reviews

No rating value

Active Filters