Skip to Content
Merck

Skip To

RTM9

Mineral Oil Rotational Viscosity Standard

viscosity 14.50 mPa.s (25.00 °C)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
500 mL
Check Cart for Availability
€ 146,00

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107

€ 146,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

14.50 mPa.s(25.00 °C)

application(s)

cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care

format

mixture

General description

This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation. Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon CCS Viscosity Standards can be found on the Paragon Scientific webpage.

Paragon ISO 17025 / ISO 17034 certified mineral oil rotational viscosity standards are specifically formulated for use with rotational viscometers.

Features and Benefits

  • Tested and certified at 20°C and 25°C.
  • Kinematic Viscosity at temperatures 20.5°C to 24.5°C is calculated in accordance with ASTM D341.
  • 500 mL inert glass packaging with tamper-evident cap provides absolute assurance of sample integrity.
  • Full traceability to national standards.
  • Supplied with the latest EU fully compliant MSDS.
  • Shelf life of 12 months.


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RTM31RTM30RTM11
format

mixture

format

mixture

format

mixture

format

mixture

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

application(s)

cosmetics
flavors and fragrances
food and beverages
oil and petrochemicals
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

cleaning products
cosmetics
cosmetics
flavors and fragrances
food and beverages
personal care

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

manufacturer/tradename

Paragon Scientific Ltd

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

viscosity

14.50 mPa.s(25.00 °C)

viscosity

5738 mPa.s(25.00 °C)

viscosity

4588 mPa.s(25.00 °C)

viscosity

29.04 mPa.s(25.00 °C)


Still not finding the right product?


signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
RTM9-500ML04061837681660

Questions

  1. Hola! Son estos patrones de viscosidad minerales con boca ancha para no tener que transvasar el producto a otro vaso y así evitar la contaminación a la hora de medir?

    1 answer
    1. These mineral oil viscosity standards are sold in narrow-mouth bottles rather than wide-mouth containers to prioritize long-term chemical stability and prevent errors during calibration. Transferring the oil to a measurement cup can introduce contamination, while wide-mouth containers present greater risks to the standard's integrity.

      Helpful?

Reviews

No rating value

Active Filters