Skip to Content
Merck
All Photos(1)

Key Documents

PHR1395

Supelco

Methocarbamol

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Guaiacol glyceryl ether carbamate, Methocarbamol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€95.80

€95.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€95.80

About This Item

Empirical Formula (Hill Notation):
C11H15NO5
CAS Number:
Molecular Weight:
241.24
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

€95.80


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1412008

API family

methocarbamol

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(OCC(O)COC1=C(OC)C=CC=C1)=O

InChI

1S/C11H15NO5/c1-15-9-4-2-3-5-10(9)16-6-8(13)7-17-11(12)14/h2-5,8,13H,6-7H2,1H3,(H2,12,14)

InChI key

GNXFOGHNGIVQEH-UHFFFAOYSA-N

Gene Information

human ... CA1(759)

Looking for similar products? Visit Product Comparison Guide

General description

Methocarbamol is drug, which is widely used as a skeletal muscle relaxant.[1] It is also used in the treatment of musculoskeletal conditions, generally associated with painful muscle spasms, cramps, and dislocations.[2]
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Methocarbamol may be used as a secondary pharma standard for the quantification of the analyte in pharmaceutical formulations[3][1] using liquid chromatography technique[3][1] and spectrophotometric technique.[3]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC4792 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of methocarbamol in equine serum and urine by high-performance liquid chromatography with ultraviolet detection and atmospheric pressure ionization-mass spectrometric confirmation
Koupai-Abyazani RM, et al.
Journal of Analytical Toxicology, 21(4), 301-305 (1997)
Validation of a RP-LC method for simultaneous determination of paracetamol, methocarbamol and diclofenac potassium in tablets
Subramanian G, et al.
Indian Journal of Pharmaceutical Sciences, 67(2), 260-260 (2005)
N Erk et al.
Journal of pharmaceutical and biomedical analysis, 24(3), 469-475 (2001-02-24)
The application of the ratio spectra derivative spectrophotometry and high-performance liquid chromatography (HPLC) to the simultaneous determination of paracetamol (PAR) and methocarbamol (MET) in combined pharmaceutical tablets is presented. The spectrophotometric procedure is based on the use of the first

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service